Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals

Have Questions?
Catalog number 167852500
Price (EUR)
50,65
Online exclusive
55,80
Save 5,15 (9%)
Each
Request bulk or custom format
Price (EUR)
50,65
Online exclusive
55,80
Save 5,15 (9%)
Each
Chemical Identifiers
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
UVat 270 nm A: =<0.5
Water=<0.1 %
Appearance (Form)Clear liquid
GC>=99.85 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only