Azidotrimethylsilane, 94%
Azidotrimethylsilane, 94%
Azidotrimethylsilane, 94%
Thermo Scientific Chemicals

Azidotrimethylsilane, 94%

CAS: 4648-54-8 | C3H9N3Si | 115.21 g/mol
数量:
10 mL
50 mL
250 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 167932500
価格(JPY)
-
数量:
250 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4648-54-8
Infrared spectrumConforms
Refractive index1.4110 to 1.4180 (20°C, 589 nm)
Appearance (Color)Clear to slightly hazy colorless
Appearance (Form)Liquid
GC>=92 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, For Azidation/Diazo transfer
RUO – Research Use Only