Methacrylic acid, 99.5%, extra pure, stabilized
Methacrylic acid, 99.5%, extra pure, stabilized
Methacrylic acid, 99.5%, extra pure, stabilized
Thermo Scientific Chemicals

Methacrylic acid, 99.5%, extra pure, stabilized

CAS: 79-41-4 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
4 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 168310010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS79-41-4
IUPAC Name2-methylprop-2-enoic acid
Molecular FormulaC4H6O2
InChI KeyCERQOIWHTDAKMF-UHFFFAOYSA-N
SMILESCC(=C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.4 %
Color scale=<20 APHA
Appearance (Form)Clear liquid
Refractive index1.4295 to 1.4315 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only