Ethyl 4-nitrobenzoate, 99%
Ethyl 4-nitrobenzoate, 99%
Ethyl 4-nitrobenzoate, 99%
Thermo Scientific Chemicals

Ethyl 4-nitrobenzoate, 99%

CAS: 99-77-4 | C9H9NO4 | 195.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 168425000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS99-77-4
IUPAC Nameethyl 4-nitrobenzoate
Molecular FormulaC9H9NO4
InChI KeyPHWSCBWNPZDYRI-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point56°C to 59°C
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only