alpha,alpha,alpha-Trifluoro-m-cresol, 98+%
alpha,alpha,alpha-Trifluoro-m-cresol, 98+%
alpha,alpha,alpha-Trifluoro-m-cresol, 98+%
Thermo Scientific Chemicals

alpha,alpha,alpha-Trifluoro-m-cresol, 98+%

CAS: 98-17-9 | C7H5F3O | 162.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
16847025025 mLGlass bottle
Catalog number 168470250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS98-17-9
IUPAC Name3-(trifluoromethyl)phenol
Molecular FormulaC7H5F3O
InChI KeyUGEJOEBBMPOJMT-UHFFFAOYSA-N
SMILESOC1=CC=CC(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
GC>=98.0 %
Refractive index1.4570 to 1.4600 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only