alpha,alpha,alpha-Trifluoro-m-cresol, 98+%
alpha,alpha,alpha-Trifluoro-m-cresol, 98+%
alpha,alpha,alpha-Trifluoro-m-cresol, 98+%
Thermo Scientific Chemicals

alpha,alpha,alpha-Trifluoro-m-cresol, 98+%

CAS: 98-17-9 | C7H5F3O | 162.11 g/mol
数量:
25 mL
100 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 168471000
価格(JPY)
-
数量:
100 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS98-17-9
IUPAC Name3-(trifluoromethyl)phenol
Molecular FormulaC7H5F3O
InChI KeyUGEJOEBBMPOJMT-UHFFFAOYSA-N
SMILESOC1=CC=CC(=C1)C(F)(F)F
さらに表示
GC>=98.0 %
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Refractive index1.4570 to 1.4600 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only