Sodium cyanoborohydride, 95%
Sodium cyanoborohydride, 95%
Sodium cyanoborohydride, 95%
Thermo Scientific Chemicals

Sodium cyanoborohydride, 95%

CAS: 25895-60-7 ∣ CH3BNNa ∣ 62.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 168550100
Price (USD)
75.60
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
75.60
Each
Chemical Identifiers
CAS25895-60-7
IUPAC Namesodium cyanoboranuide
Molecular FormulaCH3BNNa
InChI KeyCVDUGUOQTVTBJH-UHFFFAOYSA-N
SMILES[Na+].[BH3-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white to light yellow to beige
Appearance (Form)Powder
Titration Iodometric94 to 106 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

RUO – Research Use Only