Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol
Thermo Scientific Chemicals

Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol

CAS: 124-41-4 | CH3NaO | 54.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
10 L
Catalog number 168600025
Price (USD)
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS124-41-4
IUPAC Namesodium methanolate
Molecular FormulaCH3NaO
InChI KeyWQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES[Na+].C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Solution
Strength5.3 to 5.6 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only