Ammonium formate, 97%
Ammonium formate, 97%
Ammonium formate, 97%
Thermo Scientific Chemicals

Ammonium formate, 97%

CAS: 540-69-2 | CH5NO2 | 63.06 g/mol
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 168610051
Price (JPY)
-
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS540-69-2
IUPAC Nameformic acid amine
Molecular FormulaCH5NO2
InChI KeyVZTDIZULWFCMLS-UHFFFAOYSA-N
SMILESN.OC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystals
Titration with HClO496.0 to 104.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only