Phenanthrenequinone, 95%
Phenanthrenequinone, 95%
Phenanthrenequinone, 95%
Thermo Scientific Chemicals

Phenanthrenequinone, 95%

CAS: 84-11-7 | C14H8O2 | 208.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 168790250
Price (EUR)
68,60
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
68,60
Each
Chemical Identifiers
CAS84-11-7
IUPAC Name9,10-dihydrophenanthrene-9,10-dione
Molecular FormulaC14H8O2
InChI KeyYYVYAPXYZVYDHN-UHFFFAOYSA-N
SMILESO=C1C(=O)C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange-brown
Appearance (Form)Powder
Melting point203°C to 211°C
Infrared spectrumConforms
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only