3',4'-Dimethoxyacetophenone, 98%
3',4'-Dimethoxyacetophenone, 98%
3',4'-Dimethoxyacetophenone, 98%
Thermo Scientific Chemicals

3',4'-Dimethoxyacetophenone, 98%

CAS: 1131-62-0 | C10H12O3 | 180.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 168850250
Price (EUR)
34,20
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
34,20
Each
Chemical Identifiers
CAS1131-62-0
IUPAC Name1-(3,4-dimethoxyphenyl)ethan-1-one
Molecular FormulaC10H12O3
InChI KeyIQZLUWLMQNGTIW-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1OC)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Yellow to beige
Infrared spectrumConforms
GC>=97.5 %
Melting point48°C to 52°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only