3-Nitrophthalic anhydride, 97%
3-Nitrophthalic anhydride, 97%
3-Nitrophthalic anhydride, 97%
Thermo Scientific Chemicals

3-Nitrophthalic anhydride, 97%

CAS: 641-70-3 | C8H3NO5 | 193.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 168991000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS641-70-3
IUPAC Name4-nitro-1,3-dihydro-2-benzofuran-1,3-dione
Molecular FormulaC8H3NO5
InChI KeyROFZMKDROVBLNY-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige
Appearance (Form)Crystalline powder to crystals
Infrared spectrumConforms
Melting point160°C to 165°C
Titration by coupling96.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only