N-Carbobenzyloxy-L-alanine, 98%
N-Carbobenzyloxy-L-alanine, 98%
N-Carbobenzyloxy-L-alanine, 98%
Acros Organics

N-Carbobenzyloxy-L-alanine, 98%

CAS: 1142-20-7 | C11H13NO4 | 223.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 169001000
also known as 16900
HSN 29224990
Price (INR)
58,393.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
58,393.00
Each
Chemical Identifiers
CAS1142-20-7
IUPAC Name(2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid
Molecular FormulaC11H13NO4
InChI KeyTYRGLVWXHJRKMT-QMMMGPOBSA-N
SMILESC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Specific optical rotation-14° to -16° (24°C, 589 nm) (c=2, AcOH)
HPLC>=97.5 %
Appearance of solution(5% soln. in MeOH) clear, colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only