Thermo Scientific Chemicals

β-Bromostyrene, 97%, mixture of cis/trans isomers, Thermo Scientific Chemicals

Catalog number: 169290050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

β-Bromostyrene, 97%, mixture of cis/trans isomers, Thermo Scientific Chemicals

Catalog number: 169290050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 169290050
Price (USD)
Price: 33.90
Online price: 28.65
Your price:
Quantity
-

Chemical Identifiers

CAS
103-64-0
IUPAC Name
[(1E)-2-bromoethenyl]benzene
Molecular Formula
C8H7Br
InChI Key
YMOONIIMQBGTDU-VOTSOKGWSA-N
SMILES
BrC=CC1=CC=CC=C1
Appearance (Form)
Liquid
Refractive index
1.6050 to 1.6090 (20°C, 589 nm)
Appearance (Color)
Clear yellow to brown
Infrared spectrum
Conforms
GC
>=96.0 % (sum of isomers)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text