Iron(III) chloride, 98%, pure, anhydrous
Iron(III) chloride, 98%, pure, anhydrous
Iron(III) chloride, 98%, pure, anhydrous
Thermo Scientific Chemicals

Iron(III) chloride, 98%, pure, anhydrous

CAS: 7705-08-0 | Cl3Fe | 162.20 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
2.5 kg
10 kg
25 kg
Packaging:
檢視全部
Fibre drum
產品號碼 169430250
價格 (TWD)
-
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
化學識別
CAS7705-08-0
IUPAC Nameiron(3+) trichloride
Molecular FormulaCl3Fe
InChI KeyRBTARNINKXHZNM-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Fe+3]
檢視更多
規格Specification Sheet規格表
Appearance (Form)Powder
Manganese (Mn)=<3000 ppm
Arsenic (As)=<20 ppm
Zinc (Zn)=<1000 ppm
Copper (Cu)=<1000 ppm
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only