Thionyl chloride, 99.5+%, Thermo Scientific Chemicals
Thionyl chloride, 99.5+%, Thermo Scientific Chemicals
Thionyl chloride, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thionyl chloride, 99.5+%, Thermo Scientific Chemicals

数量:
25 g
100 g
500 g
1 kg
2.5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 169491000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7719-09-7
IUPAC Namesulfurooyl dichloride
Molecular FormulaCl2OS
InChI KeyFYSNRJHAOHDILO-UHFFFAOYSA-N
SMILESClS(Cl)=O
さらに表示
GC>=99.50 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC=<0.5 % Sulfuryl chloride
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only