Methyl (triphenylphosphoranylidene)acetate, 98%
Methyl (triphenylphosphoranylidene)acetate, 98%
Methyl (triphenylphosphoranylidene)acetate, 98%
Thermo Scientific Chemicals

Methyl (triphenylphosphoranylidene)acetate, 98%

CAS: 2605-67-6 | C21H19O2P | 334.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 169560250
Price (USD)
117.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
117.00
Each
Chemical Identifiers
CAS2605-67-6
IUPAC Namemethyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Molecular FormulaC21H19O2P
InChI KeyNTNUDYROPUKXNA-UHFFFAOYSA-N
SMILESCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Melting point166°C to 172°C
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only