Thermo Scientific Chemicals

Paraformaldehyde, 90%, pure, Thermo Scientific Chemicals

Catalog number: 169650010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Paraformaldehyde, 90%, pure, Thermo Scientific Chemicals

Catalog number: 169650010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 169650010
Price (USD)
Quantity
-

Chemical Identifiers

CAS
30525-89-4
IUPAC Name
formaldehyde
Molecular Formula
CH2O
InChI Key
WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES
C=O
Appearance (Color)
White
Appearance (Form)
Powder or beads or granules or prills
Infrared spectrum
Conforms
Titration with HCl
>=88 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text