4-Methylquinoline, 99%
4-Methylquinoline, 99%
4-Methylquinoline, 99%
Thermo Scientific Chemicals

4-Methylquinoline, 99%

CAS: 491-35-0 | C10H9N | 143.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 169771000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS491-35-0
IUPAC Name4-methylquinoline
Molecular FormulaC10H9N
InChI KeyMUDSDYNRBDKLGK-UHFFFAOYSA-N
SMILESCC1=C2C=CC=CC2=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to green to brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.6180 to 1.6220 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only