4-(Benzyloxy)phenol, 98%
4-(Benzyloxy)phenol, 98%
4-(Benzyloxy)phenol, 98%
Thermo Scientific Chemicals

4-(Benzyloxy)phenol, 98%

CAS: 103-16-2 | C13H12O2 | 200.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 169780010
Price (USD)
612.65
Special offer
Online exclusive
Ends: 28-Dec-2026
720.00
Save 107.35
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
612.65
Special offer
Online exclusive
Ends: 28-Dec-2026
720.00
Save 107.35
Each
Chemical Identifiers
CAS103-16-2
IUPAC Name4-(benzyloxy)phenol
Molecular FormulaC13H12O2
InChI KeyVYQNWZOUAUKGHI-UHFFFAOYSA-N
SMILESOC1=CC=C(OCC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point117°C to 122°C
Appearance (Color)Off-white to beige to brown
Appearance (Form)Crystalline powder or crystals and/or chunks
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only