Phenoxyacetic acid, 98+%
Phenoxyacetic acid, 98+%
Phenoxyacetic acid, 98+%
Thermo Scientific Chemicals

Phenoxyacetic acid, 98+%

CAS: 122-59-8 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 169870025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS122-59-8
IUPAC Name2-phenoxyacetic acid
Molecular FormulaC8H8O3
InChI KeyLCPDWSOZIOUXRV-UHFFFAOYSA-N
SMILESOC(=O)COC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.0 %
Melting point98°C to 102°C
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only