Thermo Scientific Chemicals

Phenoxyacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 169871000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Phenoxyacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 169871000
100 g, Plastic bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
122-59-8
Titration with NaOH
>=98.0 %
Melting point
98°C to 102°C
Appearance (Color)
White to beige
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text