Nunc™ Non-treated Flasks
Thermo Scientific™
Nunc™ Non-treated Flasks
Minimize the chance of contamination with these high-quality, optically clear polystyrene Nunc Non-Treated Flasks for suspension cell cultures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCulture Area
16990025 cm²
132903500 cm²
15680075 cm²
159926175 cm²
Catalog number 169900
Price (USD)
587.65
Online exclusive
646.00
Save 58.35
Case of 200
Culture Area:
25 cm²
Price (USD)
587.65
Online exclusive
646.00
Save 58.35
Case of 200
Benefit from the user-friendly designs of these high-quality, optically clear polystyrene Thermo Scientific Nunc Non-Treated Flasks for suspension cell cultures.
Features
  • White caps for easy identification
  • Ergonomic design for easy handling and pipetting
  • TripleFlask with three parallel layers has the same external dimensions of a standard 175 cm2 flask
  • Filter cap ensures constant airflow while minimizing chances of contamination
  • Sterilie and non-pyrogenic
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorWhite
Closure TypeFilter
DescriptionNunc Non-treated T25 EasYFlask, Filter Cap
Neck StyleAngled
No. per Case200
No. per Pack10
Volume (Metric) Working7 mL
Bottom ShapeFlat
Culture Area25 cm²
MaterialPolystyrene
SterilitySterile
Surface TreatmentUntreated
Unit SizeCase of 200