Thermo Scientific™
Samco™ 17 × 100mm Disposable Culture Tubes (DCTs)
Catalog number: 17-001S
Thermo Scientific™
Samco™ 17 × 100mm Disposable Culture Tubes (DCTs)
Catalog number: 17-001S
Perform most lab applications with Thermo Scientific™ Samco™ 17 × 100mm Disposable Culture Tubes, which deliver lot-to-lot uniformity and reliable performance. These chemical-free and biologically inert tubes are offered in clarified polypropylene for strength and resistance, or polystyrene for superior clarity. Each tube is precision molded and manufactured from premium grade materials by processes monitored with quality checks.
Catalog Number
17-001S
Unit Size
Each
Description
Tubes only, no threads
Material
Polypropylene
Closure Type
Snap Cap
Price (USD)
Availability
***
Full specifications
Quantity
-
DescriptionTubes only, no threads
Closure TypeSnap Cap
Length (Metric)100 mm
RCF Rating3000 RCF
MaterialPolypropylene
No. per Pack125
No. per Case1000
SterilitySterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeEach
Catalog Number
Unit Size
Case of 500
Description
Tubes and snap caps, pre-assembled, blister tray
Material
Polypropylene
Closure Type
Snap Cap
Price (USD)
Availability
***
Full specifications
Quantity
-
Closure ColorBlue
DescriptionTubes and snap caps, pre-assembled, blister tray
Closure TypeSnap Cap
ColorNatural
Length (Metric)100 mm
RCF Rating3000 RCF
MaterialPolypropylene
No. per Pack25
No. per Case500
SterilitySterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeCase of 500
Catalog Number
Unit Size
Each
Description
ubes and snap caps, pre-assembled, individually wrapped
Material
Polypropylene
Closure Type
Snap Cap
Price (USD)
Availability
***
Full specifications
Quantity
-
Closure ColorNatural
Descriptionubes and snap caps, pre-assembled, individually wrapped
Closure TypeSnap Cap
Length (Metric)100 mm
RCF Rating3000 RCF
MaterialPolypropylene
No. per Pack1
No. per Case500
SterilitySterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Tubes and snap caps, pre-assembled
Material
Polystyrene
Closure Type
Snap Cap
Price (USD)
Availability
***
Full specifications
Quantity
-
Closure ColorNatural
DescriptionTubes and snap caps, pre-assembled
Closure TypeSnap Cap
ColorNatural
Length (Metric)100 mm
RCF Rating10000
MaterialPolystyrene
No. per Pack125
No. per Case1000
SterilitySterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeEach
Catalog Number
Unit Size
Case of,500
Description
Tubes and snap caps, pre-assembled
Material
Polystyrene
Closure Type
Snap Cap
Price (USD)
Availability
***
Full specifications
Quantity
-
Closure ColorBlue
DescriptionTubes and snap caps, pre-assembled
Closure TypeSnap Cap
ColorNatural
Length (Metric)100 mm
RCF Rating10000
MaterialPolystyrene
No. per Pack25
No. per Case500
SterilitySterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeCase of,500
Catalog Number
Unit Size
Case of 1000
Description
Tubes only, no threads, bulk
Material
Polypropylene
Closure Type
Snap Cap
Price (USD)
239.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionTubes only, no threads, bulk
Closure TypeSnap Cap
Length (Metric)100 mm
RCF Rating3000 RCF
MaterialPolypropylene
No. per Pack1000
No. per Case1000
SterilityNon-sterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeCase of 1000
Catalog Number
Unit Size
Each
Description
Tubes only, no threads, bulk
Material
Polystyrene
Closure Type
Snap Cap
Price (USD)
Availability
***
Full specifications
Quantity
-
DescriptionTubes only, no threads, bulk
Closure TypeSnap Cap
Length (Metric)100 mm
RCF Rating3000 RCF
MaterialPolystyrene
No. per Pack1000
No. per Case1000
SterilityNon-sterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Tubes only, no threads, bulk
Material
Polypropylene
Closure Type
Snap Cap
Price (USD)
Availability
***
Full specifications
Quantity
-
DescriptionTubes only, no threads, bulk
Closure TypeSnap Cap
Length (Metric)100 mm
RCF Rating3000 RCF
MaterialPolypropylene
No. per Pack1000
No. per Case1000
SterilityNon-sterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeEach
Showing 8 of 8
Catalog NumberSpecificationsUnit SizeDescriptionMaterialClosure TypePrice (USD)AvailabilityQuantity
17-001SFull specifications
EachTubes only, no threadsPolypropyleneSnap CapRequest A Quote***-
DescriptionTubes only, no threads
Closure TypeSnap Cap
Length (Metric)100 mm
RCF Rating3000 RCF
MaterialPolypropylene
No. per Pack125
No. per Case1000
SterilitySterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeEach
17-003SFull specifications
Case of 500Tubes and snap caps, pre-assembled, blister trayPolypropyleneSnap CapRequest A Quote***-
Closure ColorBlue
DescriptionTubes and snap caps, pre-assembled, blister tray
Closure TypeSnap Cap
ColorNatural
Length (Metric)100 mm
RCF Rating3000 RCF
MaterialPolypropylene
No. per Pack25
No. per Case500
SterilitySterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeCase of 500
17-005SFull specifications
Eachubes and snap caps, pre-assembled, individually wrappedPolypropyleneSnap CapRequest A Quote***-
Closure ColorNatural
Descriptionubes and snap caps, pre-assembled, individually wrapped
Closure TypeSnap Cap
Length (Metric)100 mm
RCF Rating3000 RCF
MaterialPolypropylene
No. per Pack1
No. per Case500
SterilitySterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeEach
17-006SFull specifications
EachTubes and snap caps, pre-assembledPolystyreneSnap CapRequest A Quote***-
Closure ColorNatural
DescriptionTubes and snap caps, pre-assembled
Closure TypeSnap Cap
ColorNatural
Length (Metric)100 mm
RCF Rating10000
MaterialPolystyrene
No. per Pack125
No. per Case1000
SterilitySterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeEach
17-009SFull specifications
Case of,500Tubes and snap caps, pre-assembledPolystyreneSnap CapRequest A Quote***-
Closure ColorBlue
DescriptionTubes and snap caps, pre-assembled
Closure TypeSnap Cap
ColorNatural
Length (Metric)100 mm
RCF Rating10000
MaterialPolystyrene
No. per Pack25
No. per Case500
SterilitySterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeCase of,500
17-010Full specifications
Case of 1000Tubes only, no threads, bulkPolypropyleneSnap Cap
239.00
Your Price:
***
DescriptionTubes only, no threads, bulk
Closure TypeSnap Cap
Length (Metric)100 mm
RCF Rating3000 RCF
MaterialPolypropylene
No. per Pack1000
No. per Case1000
SterilityNon-sterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeCase of 1000
17-0011Full specifications
EachTubes only, no threads, bulkPolystyreneSnap CapRequest A Quote***-
DescriptionTubes only, no threads, bulk
Closure TypeSnap Cap
Length (Metric)100 mm
RCF Rating3000 RCF
MaterialPolystyrene
No. per Pack1000
No. per Case1000
SterilityNon-sterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeEach
17-0010Full specifications
EachTubes only, no threads, bulkPolypropyleneSnap CapRequest A Quote***-
DescriptionTubes only, no threads, bulk
Closure TypeSnap Cap
Length (Metric)100 mm
RCF Rating3000 RCF
MaterialPolypropylene
No. per Pack1000
No. per Case1000
SterilityNon-sterile
Bulk PackYes
Diameter (Metric) Outer17 mm
TypeCulture Tube
ShapeSquare
Unit SizeEach
Showing 8 of 8
Note:
Available with a two-position snap cap or without caps.
Sterile Tubes:
  • Gamma irradiated
  • Ready to use
  • Bulk or individually wrapped
Non-Sterile Tubes:
  • Bulk or wrapped in packs
Figures