eBioscience™ Streptavidin APC Conjugate
Invitrogen™

eBioscience™ Streptavidin APC Conjugate

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Have Questions?
產品號碼Quantity
17-4317-82100 μg
產品號碼 17-4317-82
價格 (TWD)
6,144.00
線上優惠
Ends: 30-Sep-2026
7,680.00
您節省 1,536.00 (20%)
Each
Quantity:
100 μg
價格 (TWD)
6,144.00
線上優惠
Ends: 30-Sep-2026
7,680.00
您節省 1,536.00 (20%)
Each

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser

Red Laser

Emit

660 nm

Excite

633 - 647 nm

Reported Application

Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
規格
Concentration0.2 mg/mL
Label or DyeAPC (Allophycocyanin)
Product TypeStreptavidin Conjugate (Fluorescent)
Quantity100 μg
Target MoleculeBiotin
ConjugateAPC
FormLiquid
Product LineeBioscience
Unit SizeEach