Methyl p-toluenesulfonate, 97%
Methyl p-toluenesulfonate, 97%
Methyl p-toluenesulfonate, 97%
Thermo Scientific Chemicals

Methyl p-toluenesulfonate, 97%

CAS: 80-48-8 | C8H10O3S | 186.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 170100025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS80-48-8
IUPAC Namemethyl 4-methylbenzene-1-sulfonate
Molecular FormulaC8H10O3S
InChI KeyVUQUOGPMUUJORT-UHFFFAOYSA-N
SMILESCOS(=O)(=O)C1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Infrared spectrumConforms
Melting point24°C to 30°C
Titration excess NaOH96.0 to 104.0 %
Water=<0.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only