2-Bromo-4'-methylacetophenone, 97%
2-Bromo-4'-methylacetophenone, 97%
2-Bromo-4'-methylacetophenone, 97%
Thermo Scientific Chemicals

2-Bromo-4'-methylacetophenone, 97%

CAS: 619-41-0 | C9H9BrO | 213.07 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
檢視全部
Plastic bottle
產品號碼 170390250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS619-41-0
IUPAC Name2-bromo-1-(4-methylphenyl)ethan-1-one
Molecular FormulaC9H9BrO
InChI KeyKRVGXFREOJHJAX-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(=O)CBr
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only