2-Furonitrile, 99%
2-Furonitrile, 99%
2-Furonitrile, 99%
Thermo Scientific Chemicals

2-Furonitrile, 99%

CAS: 617-90-3 | C5H3NO | 93.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 170430250
Price (EUR)
208,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
208,00
Each
Chemical Identifiers
CAS617-90-3
IUPAC Namefuran-2-carbonitrile
Molecular FormulaC5H3NO
InChI KeyYXDXXGXWFJCXEB-UHFFFAOYSA-N
SMILESN#CC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to light orange
AppearanceMay darken on storage
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4780 to 1.4800 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only