You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Facility Safety and Maintenance
▸
Signs and Tags
▸
Facilities Maintenance Signs
▸
Maximum Occupancy People Sign
Actual product may vary
Maximum Occupancy People Sign
Post this sign to keep employees and visitors informed on the best way to keep safe social distances.
Have Questions?
Catalog Number
Dimensions (L x W)
Material
170615
10 x 14 in.
Aluminum
Catalog number
170615
HSN
39233090
Price (INR)
13,474.00
CS
Add to cart
Dimensions (L x W):
10 x 14 in.
Material:
Aluminum
Price (INR)
13,474.00
CS
Add to cart
This B-401 polystyrene material is ideal for use in light duty areas and is perfect for temporary identification
With this temporary sign, you get excellent abrasion and chemical resistance so your identification remains impactful in harsh conditions
Great for indoor and outdoor use
Service temperature of -40 to 175°F (-40 to 80°C)
Durable non-reflective aluminum material (B-555) provides lasting identification with durable inks and clear-coat protection for added fade resistance
Designed for both indoor and outdoor use
Withstands wind, rain, sunlight and high temperatures
Specifications
Material
Aluminum
Mounting Type
Corner Holes
Languages
English
Dimensions (L x W)
10 x 14 in.
Displayed Text
MAXIMUM OCCUPANCY __ PEOPLE
Color
Blue on White
Unit Size
CS