1-(4-Methoxyphenyl)-1-cyclopropanecarbonitrile, 96%
1-(4-Methoxyphenyl)-1-cyclopropanecarbonitrile, 96%
1-(4-Methoxyphenyl)-1-cyclopropanecarbonitrile, 96%
Thermo Scientific Chemicals

1-(4-Methoxyphenyl)-1-cyclopropanecarbonitrile, 96%

CAS: 16728-00-0 | C11H11NO | 173.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
17062025025 gGlass bottle
Catalog number 170620250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16728-00-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5415 to 1.5435 (20°C, 589 nm)
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only