4,4'-Bis(diethylamino)benzophenone, 99+%
4,4'-Bis(diethylamino)benzophenone, 99+%
4,4'-Bis(diethylamino)benzophenone, 99+%
4,4'-Bis(diethylamino)benzophenone, 99+%
Thermo Scientific Chemicals

4,4'-Bis(diethylamino)benzophenone, 99+%

CAS: 90-93-7 | C21H28N2O | 324.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 170811000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS90-93-7
IUPAC Name4-[4-(diethylamino)benzoyl]-N,N-diethylaniline
Molecular FormulaC21H28N2O
InChI KeyVYHBFRJRBHMIQZ-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to yellow-green
Appearance (Form)Powder or granules and/or lumps
Infrared spectrumConforms
Melting point93°C to 96°C
HPLC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only