Phthalimide, potassium derivative, 99%
Phthalimide, potassium derivative, 99%
Phthalimide, potassium derivative, 99%
Thermo Scientific Chemicals

Phthalimide, potassium derivative, 99%

CAS: 1074-82-4 | C8H4KNO2 | 185.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 170865000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1074-82-4
IUPAC Namepotassium 1,3-dioxo-2,3-dihydro-1H-isoindol-2-ide
Molecular FormulaC8H4KNO2
InChI KeyFYRHIOVKTDQVFC-UHFFFAOYSA-M
SMILES[K+].O=C1[N-]C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=98.5 % (On dried substance)
Appearance (Color)White to yellow to green
Water=<0.5 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only