Ethidium bromide, 95%, pure
Ethidium bromide, 95%, pure
Ethidium bromide, 95%, pure
Thermo Scientific Chemicals

Ethidium bromide, 95%, pure

CAS: 1239-45-8 | C21H20BrN3 | 394.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 170960050
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Chemical Identifiers
CAS1239-45-8
IUPAC Name3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
Molecular FormulaC21H20BrN3
InChI KeyZMMJGEGLRURXTF-UHFFFAOYSA-N
SMILES[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to dark purple
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration Argentometric>=94 %
Loss on drying=<5.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only