Succinonitrile, 99+%
Succinonitrile, 99+%
Succinonitrile, 99+%
Thermo Scientific Chemicals

Succinonitrile, 99+%

CAS: 110-61-2 | C4H4N2 | 80.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 171185000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS110-61-2
IUPAC Namebutanedinitrile
Molecular FormulaC4H4N2
InChI KeyIAHFWCOBPZCAEA-UHFFFAOYSA-N
SMILESN#CCCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)Colorless to white to light yellow
Appearance (Form)Crystalline mass or waxy solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only