Paraffin, liquid, pure
Paraffin, liquid, pure
Paraffin, liquid, pure
Thermo Scientific Chemicals

Paraffin, liquid, pure

CAS: 8012-95-1 | MFCD00131611
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 171400025
Price (EUR)
94,65
Online exclusive
105,00
Save 10,35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
94,65
Online exclusive
105,00
Save 10,35
Each
Chemical Identifiers
CAS8012-95-1
Molecular FormulaMFCD00131611
SpecificationsSpecification SheetSpecification Sheet
Viscosity63 to 80 mm²/s (40°C)
Appearance (Form)Oily liquid
Infrared spectrumConforms
Refractive index1.4700 to 1.4800 (20°C, 589 nm)
Density(20°C) 0.827 to 0.890 g/ml
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only