1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%
Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%

CAS: 25952-53-8 | C8H18ClN3 | 191.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 171445000
Price (USD)
2,516.65
Special offer
Online exclusive
Ends: 28-Dec-2026
2,960.00
Save 443.35
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
2,516.65
Special offer
Online exclusive
Ends: 28-Dec-2026
2,960.00
Save 443.35
Each
Chemical Identifiers
CAS25952-53-8
IUPAC Name({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine hydrochloride
Molecular FormulaC8H18ClN3
InChI KeyFPQQSJJWHUJYPU-UHFFFAOYSA-N
SMILESCl.CCN=C=NCCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point111°C to 116°C
Titration Argentometric>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only