alpha,alpha-Diphenylglycine, 98%
alpha,alpha-Diphenylglycine, 98%
alpha,alpha-Diphenylglycine, 98%
Thermo Scientific Chemicals

alpha,alpha-Diphenylglycine, 98%

CAS: 3060-50-2 | C14H13NO2 | 227.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 171720250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Assay Percent Range98%
Beilstein14,I,625
CAS3060-50-2
Chemical Name or Materialα,α-Diphenylglycine
ColorWhite
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only