alpha,alpha-Diphenylglycine, 98%
alpha,alpha-Diphenylglycine, 98%
alpha,alpha-Diphenylglycine, 98%
Acros Organics

alpha,alpha-Diphenylglycine, 98%

CAS: 3060-50-2 | C14H13NO2 | 227.26 g/mol
Have Questions?
Catalog NumberQuantityPackaging
171721000
also known as 17172
100 gGlass bottle
Catalog number 171721000
also known as 17172
HSN 32049000
Price (INR)
207,376.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
207,376.00
Each
Chemical Identifiers
CAS3060-50-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
Titration with TBAH97.5 to 102.5 %
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only