1,3-Diethyl-2-thiobarbituric acid, 99%
1,3-Diethyl-2-thiobarbituric acid, 99%
1,3-Diethyl-2-thiobarbituric acid, 99%
1,3-Diethyl-2-thiobarbituric acid, 99%
Thermo Scientific Chemicals

1,3-Diethyl-2-thiobarbituric acid, 99%

CAS: 5217-47-0 | C8H12N2O2S | 200.26 g/mol
数量:
5 g
50 g
250 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 171742500
価格(JPY)
-
見積もりを依頼する
数量:
250 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5217-47-0
IUPAC Name1,3-diethyl-2-sulfanylidene-1,3-diazinane-4,6-dione
Molecular FormulaC8H12N2O2S
InChI KeySHBTUGJAKBRBBJ-UHFFFAOYSA-N
SMILESCCN1C(=O)CC(=O)N(CC)C1=S
さらに表示
GC>=98.5 %
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point105°C to 109°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only