N-Isopropylcyclohexylamine, 99%
N-Isopropylcyclohexylamine, 99%
N-Isopropylcyclohexylamine, 99%
N-Isopropylcyclohexylamine, 99%
Thermo Scientific Chemicals

N-Isopropylcyclohexylamine, 99%

CAS: 1195-42-2 | C9H19N | 141.26 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 171761000
価格(JPY)
-
見積もりを依頼する
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1195-42-2
IUPAC NameN-(propan-2-yl)cyclohexanamine
Molecular FormulaC9H19N
InChI KeyUYYCVBASZNFFRX-UHFFFAOYSA-N
SMILESCC(C)NC1CCCCC1
さらに表示
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Refractive index1.4470 to 1.4490 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only