p-Toluenesulfonic acid monohydrate, 97.5%, pure
p-Toluenesulfonic acid monohydrate, 97.5%, pure
p-Toluenesulfonic acid monohydrate, 97.5%, pure
Thermo Scientific Chemicals

p-Toluenesulfonic acid monohydrate, 97.5%, pure

CAS: 6192-52-5 | C7H8O3S·H2O | 190.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 171785000
Price (USD)
40.65
Online exclusive
44.60
Save 3.95
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
40.65
Online exclusive
44.60
Save 3.95
Each
Chemical Identifiers
CAS6192-52-5
IUPAC Name4-methylbenzene-1-sulfonic acid hydrate
Molecular FormulaC7H10O4S
InChI KeyKJIFKLIQANRMOU-UHFFFAOYSA-N
SMILESO.CC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Adhering crystals
AppearanceMay discolor to pink on storage
Infrared spectrumConforms
Titration with NaOH>=96.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only