4-Fluorobenzylamine, 97%
4-Fluorobenzylamine, 97%
4-Fluorobenzylamine, 97%
Thermo Scientific Chemicals

4-Fluorobenzylamine, 97%

CAS: 140-75-0 | C7H8FN | 125.15 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
產品號碼 171951000
價格 (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS140-75-0
IUPAC Name1-(4-fluorophenyl)methanamine
Molecular FormulaC7H8FN
InChI KeyIIFVWLUQBAIPMJ-UHFFFAOYSA-N
SMILESNCC1=CC=C(F)C=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.5115 to 1.5155 (20°C, 589 nm)
GC>=96.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only