DL-Leucine, 99+%
DL-Leucine, 99+%
DL-Leucine, 99+%
Thermo Scientific Chemicals

DL-Leucine, 99+%

CAS: 328-39-2 | C6H13NO2 | 131.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
Packaging:
Plastic Bottle
Catalog number 172130250
Price (EUR)
33,65
Online exclusive
37,60
Save 3,95
Each
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (EUR)
33,65
Online exclusive
37,60
Save 3,95
Each
Chemical Identifiers
CAS328-39-2
IUPAC Name2-amino-4-methylpentanoic acid
Molecular FormulaC6H13NO2
InChI KeyROHFNLRQFUQHCH-UHFFFAOYNA-N
SMILESCC(C)CC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<20 ppm
Sulfated ash=<0.1 %
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only