2-[2-(2-Chloroethoxy)ethoxy]ethanol, 95%
2-[2-(2-Chloroethoxy)ethoxy]ethanol, 95%
2-[2-(2-Chloroethoxy)ethoxy]ethanol, 95%
Thermo Scientific Chemicals

2-[2-(2-Chloroethoxy)ethoxy]ethanol, 95%

CAS: 5197-62-6 | C6H13ClO3 | 168.62 g/mol
数量:
10 g
50 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 172270500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5197-62-6
IUPAC Name2-[2-(2-chloroethoxy)ethoxy]ethan-1-ol
Molecular FormulaC6H13ClO3
InChI KeyKECMLGZOQMJIBM-UHFFFAOYSA-N
SMILESOCCOCCOCCCl
さらに表示
GC>=94 %
Infrared spectrumConforms
Appearance (Color)Clear light yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only