7-Methoxy-1-tetralone, 97%
7-Methoxy-1-tetralone, 97%
7-Methoxy-1-tetralone, 97%
Acros Organics

7-Methoxy-1-tetralone, 97%

CAS: 6836-19-7 | C11H12O2 | 176.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 172430050
also known as 17243
HSN 29143990
Price (INR)
9,406.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,406.00
Each
Chemical Identifiers
CAS6836-19-7
IUPAC Name7-methoxy-1,2,3,4-tetrahydronaphthalen-1-one
Molecular FormulaC11H12O2
InChI KeyGABLTKRIYDNDIN-UHFFFAOYSA-N
SMILESCOC1=CC=C2CCCC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point57°C to 64°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only