EPPS, 99%, for biochemistry
EPPS, 99%, for biochemistry
EPPS, 99%, for biochemistry
Thermo Scientific Chemicals

EPPS, 99%, for biochemistry

CAS: 16052-06-5 | C9H20N2O4S | 252.329 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 172601000
Price (EUR)
71,65
Online exclusive
79,90
Save 8,25
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
71,65
Online exclusive
79,90
Save 8,25
Each
Chemical Identifiers
CAS16052-06-5
IUPAC Name3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
Molecular FormulaC9H20N2O4S
InChI KeyOWXMKDGYPWMGEB-UHFFFAOYSA-N
SMILESOCCN1CCN(CCCS(O)(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
UV(0.1 M in water) 1 cm cell
Appearance (Color)White
Appearance (Form)Crystalline powder
pH5 to 6 (10 % w/v at 20°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only