PIPES, 98,5+ %, para bioquímica
PIPES, 98,5+ %, para bioquímica
PIPES, 98,5+ %, para bioquímica
Thermo Scientific Chemicals

PIPES, 98,5+ %, para bioquímica

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
500 g
2.5 kg
Envase:
Frasco de plástico
Número de catálogo 172615000
Precio (EUR)
367,00
Each
Cantidad:
500 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Precio (EUR)
367,00
Each
Identificadores químicos
CAS5625-37-6
IUPAC Namesodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular FormulaC8H17N2NaO6S2
InChI KeyOGGAIRCLBMGXCZ-UHFFFAOYSA-M
SMILES[Na+].OS(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
Loss on drying=<0.5 %
UV(0.1 M in 0.1 M NaOH) 1 cm cell
UVat 280 nm A: =<0.04
Appearance (Color)White
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only