MOPS, 99%, for biochemistry
MOPS, 99%, for biochemistry
MOPS, 99%, for biochemistry
Thermo Scientific Chemicals

MOPS, 99%, for biochemistry

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 172630025
Price (EUR)
1.095,00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
1.095,00
Each
Chemical Identifiers
CAS1132-61-2
IUPAC Name4-(3-sulfonatopropyl)morpholin-4-ium
Molecular FormulaC7H15NO4S
InChI KeyDVLFYONBTKHTER-UHFFFAOYSA-N
SMILES[O-]S(=O)(=O)CCC[NH+]1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 260 nm A: =<0.05
Appearance (Form)Crystalline powder
Titration with NaOH>=98.5 %
pH3.0 to 4.5 (1 % in water at 25°C)
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only