Bicine, 99+%, for analysis
Bicine, 99+%, for analysis
Bicine, 99+%, for analysis
Thermo Scientific Chemicals

Bicine, 99+%, for analysis

CAS: 150-25-4 | C6H12NNaO4 | 185.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 172650010
Price (USD)
473.65
Special offer
Online exclusive
Ends: 28-Dec-2026
557.00
Save 83.35
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
473.65
Special offer
Online exclusive
Ends: 28-Dec-2026
557.00
Save 83.35
Each
Chemical Identifiers
CAS150-25-4
IUPAC Namesodium 2-[bis(2-hydroxyethyl)amino]acetate
Molecular FormulaC6H12NNaO4
InChI KeyMFBDBXAVPLFMNJ-UHFFFAOYSA-M
SMILES[Na+].OCCN(CCO)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HClO4>=99.0 %
Sulfated ash=<0.1 %
Sulfate (SO4)=<50 ppm
Heavy metals (as Pb)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only