Diaminomaleonitrile, 98%
Diaminomaleonitrile, 98%
Diaminomaleonitrile, 98%
Thermo Scientific Chemicals

Diaminomaleonitrile, 98%

CAS: 1187-42-4 | C4H4N4 | 108.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 172701000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1187-42-4
IUPAC Name(2Z)-diaminobut-2-enedinitrile
Molecular FormulaC4H4N4
InChI KeyDPZSNGJNFHWQDC-ARJAWSKDSA-N
SMILESN\C(C#N)=C(/N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
UV spectrophotometry>=97.5 %
Appearance (Color)Brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only